Dichlorodiisopropylsilane ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D2603-25G D2603-5G
TCIAD2603-25GEA 632 DKK
TCIAD2603-25G TCIAD2603-5G
Dichlorodiisopropylsilane ≥98.0% (by GC)
Dichlorodiisopropylsilane
Formel: C₆H₁₄Cl₂Si
Kogepunkt: 170°C
Flammepunkt: 43°C
MDL Number: MFCD00054895
CAS-nummer: 7751-38-4
UN nummer: 2985
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR