4-(Dimethylamino)phenyldiphenylphosphine ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D2478-1G D2478-5G
TCIAD2478-1GEA 255 DKK
TCIAD2478-1G TCIAD2478-5G
4-(Dimethylamino)phenyldiphenylphosphine ≥95.0% (by GC)
4-(Dimethylamino)phenyldiphenylphosphine
Formel: C₂₀H₂₀NP
MDL Number: MFCD00192068
CAS-nummer: 739-58-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 149.0 to 153.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR