Diethylenetriaminepentaacetic dianhydride ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D2424-25G
D2424-5G
TCIAD2424-25GEA
2380
DKK
TCIAD2424-25G
TCIAD2424-5G
Diethylenetriaminepentaacetic dianhydride ≥98.0% (by titrimetric analysis)
Diethylenetriaminepentaacetic dianhydride
Formel:
C₁₄H₁₉N₃O₈ Smeltepunkt: 182°C |
MDL Number:
MFCD00010697 CAS-nummer: 23911-26-4 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Methanolysis Method) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...