Diphenyl-1-pyrenylphosphine ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
DPPP, Diphenyl(pyren-1-yl)phosphine, 1-(Diphenylphosphino)pyrene
D2404-1G
D2404-100MG
TCIAD2404-1GEA
2430
DKK
TCIAD2404-1G
TCIAD2404-100MG
Diphenyl-1-pyrenylphosphine ≥95.0% (by GC)
Diphenyl-1-pyrenylphosphine
Formel:
C₂₈H₁₉P Molekylvægt: 386.43 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00278755 CAS-nummer: 110231-30-6 |
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 174.0 to 178.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...