2,4-Diethylthioxanthen-9-one ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2,4-Diethyl-9H-thioxanthen-9-one

D2375-25G D2375-250G
TCIAD2375-25GEA 255 DKK
TCIAD2375-25G TCIAD2375-250G
2,4-Diethylthioxanthen-9-one ≥98.0% (by GC)
2,4-Diethylthioxanthen-9-one
Formel: C₁₇H₁₆OS
MDL Number: MFCD00082910
CAS-nummer: 82799-44-8
EINECS: 280-041-0

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR