Dichlorodecylmethylsilane ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
D2348-25ML
D2348-5ML
TCIAD2348-25MLEA
567
DKK
TCIAD2348-25ML
TCIAD2348-5ML
Dichlorodecylmethylsilane ≥95.0% (by GC)
Dichlorodecylmethylsilane
Formel:
C₁₁H₂₄Cl₂Si Kogepunkt: 129°C Flammepunkt: 120°C |
MDL Number:
MFCD00053194 CAS-nummer: 18051-88-2 EINECS: 241-962-3 UN nummer: 2987 ADR: 8,II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...