3,5-Dibenzyloxybenzoic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3,5-Bis(benzyloxy)benzoic acid
D2267-25G
D2267-5G
TCIAD2267-25GEA
2700
DKK
TCIAD2267-25G
TCIAD2267-5G
3,5-Dibenzyloxybenzoic acid ≥98.0% (by titrimetric analysis)
3,5-Dibenzyloxybenzoic acid
Formel:
C₂₁H₁₈O₄ |
MDL Number:
MFCD00191341 CAS-nummer: 28917-43-3 EINECS: 249-311-5 |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 212.0 to 216.0 deg-C |
Solubility in N,N-DMF | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...