4-Dimethylamino-4'-nitrobiphenyl ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D2263-1G D2263-5G D2263-200MG
TCIAD2263-1GEA 2890 DKK
TCIAD2263-1G TCIAD2263-5G TCIAD2263-200MG
4-Dimethylamino-4'-nitrobiphenyl ≥98.0% (by HPLC)
4-Dimethylamino-4'-nitrobiphenyl
Formel: C₁₄H₁₄N₂O₂
Molekylvægt: 242.28 g/mol
MDL Number: MFCD00191632
CAS-nummer: 2143-87-5

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 243.0 to 249.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR