Dodecamethylcyclohexasiloxane ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D2040-5ML D2040-1ML D2040-25ML
TCIAD2040-5MLEA 1720 DKK
TCIAD2040-5ML TCIAD2040-1ML TCIAD2040-25ML
Dodecamethylcyclohexasiloxane ≥97.0% (by GC)
Dodecamethylcyclohexasiloxane
Formel: C₁₂H₃₆O₆Si₆
Kogepunkt: 245°C
Flammepunkt: 105°C
MDL Number: MFCD00144215
CAS-nummer: 540-97-6
EINECS: 208-762-8
Merck Index: 14,03403
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e1c1df697dfb2d8409fc6a33eda2495e82d20cd&vid=7e1c1df697dfb2d8409fc6a33eda2495e82d20cd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR