6-Dimethylamino-1-hexanol ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 6-(Dimethylamino)hexan-1-ol

D1664-25ML D1664-5ML
TCIAD1664-25MLEA 854 DKK
TCIAD1664-25ML TCIAD1664-5ML
6-Dimethylamino-1-hexanol ≥97.0% (by GC)
6-Dimethylamino-1-hexanol
Formel: C₈H₁₉NO
Kogepunkt: 117°C
MDL Number: MFCD00046028
CAS-nummer: 1862-07-3
EINECS: 404-680-3
UN nummer: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d709e4eec72c1c8d97202efd88d5a72f439361e0&vid=d709e4eec72c1c8d97202efd88d5a72f439361e0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR