4,4'-Dipyridyl sulphide ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4,4'-Thiodipyridine, Di(pyridin-4-yl)sulphane

D1483-5G D1483-1G
TCIAD1483-5GEA 2260 DKK
TCIAD1483-5G TCIAD1483-1G
4,4'-Dipyridyl sulphide ≥98.0% (by GC, titration analysis)
4,4'-Dipyridyl sulphide
Formel: C₁₀H₈N₂S
Molekylvægt: 188.25 g/mol
MDL Number: MFCD00059784
CAS-nummer: 37968-97-1

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 66.0 to 71.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR