Diphenylsulphone-3,3'-disulphonic acid disodium salt ≥97.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Disodium Diphenylsulphone-3,3'-disulphonate
D0916-25G
D0916-5G
TCIAD0916-25GEA
4190
DKK
TCIAD0916-25G
TCIAD0916-5G
Diphenylsulphone-3,3'-disulphonic acid disodium salt ≥97.0% (by HPLC, titration analysis)
Diphenylsulphone-3,3'-disulphonic acid disodium salt
Formel:
C₁₂H₁₀O₈S₃ Molekylvægt: 422.35 g/mol |
MDL Number:
MFCD00059845 CAS-nummer: 39616-93-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Ion exchange titration) | min. 97.0 % |
Solubility in Water | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...