Sodium dimethyldithiocarbamate dihydrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Sodium Dimethyldithiocarbamate Dihydrate, Dimethyldithiocarbamic acid sodium salt dihydrate

D0716-25G D0716-500G
TCIAD0716-25GEA 132 DKK
TCIAD0716-25G TCIAD0716-500G
Sodium dimethyldithiocarbamate dihydrate ≥98.0% (by titrimetric analysis)
Sodium dimethyldithiocarbamate dihydrate
Formel: C₃H₁₀NNaO₂S₂
Molekylvægt: 179.24 g/mol
MDL Number: MFCD00150729
CAS-nummer: 128-04-1
EINECS: 204-876-7
UN nummer: 3077
ADR: 9 III

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR