m-Xylene-4-sulphonic acid hydrate ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2,4-Dimethylbenzenesulphonic acid
D0680-25G
D0680-500G
TCIAD0680-25GEA
132
DKK
TCIAD0680-25G
TCIAD0680-500G
m-Xylene-4-sulphonic acid hydrate ≥98.0% (by HPLC, titration analysis)
m-Xylene-4-sulphonic acid hydrate
Formel:
C₈H₁₀O₃S |
MDL Number:
MFCD00007483 CAS-nummer: 88-61-9 EINECS: 201-843-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Melting point | 58.0 to 64.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...