Diisobutyl disulphide ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Isobutyl Disulphide
D0616-25ML
D0616-5ML
TCIAD0616-25MLEA
1350
DKK
TCIAD0616-25ML
TCIAD0616-5ML
Diisobutyl disulphide ≥98.0% (by GC)
Diisobutyl disulphide
Formel:
C₈H₁₈S₂ Kogepunkt: 109°C |
MDL Number:
MFCD00039850 CAS-nummer: 1518-72-5 EINECS: 216-177-4 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...