Zinc diethyldithiocarbamate ≥99.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Zinc dethyldithiocarbamate, Diethyldithiocarbamic Acid Zinc Salt
D0492-25G
D0492-500G
TCIAD0492-25GEA
132
DKK
TCIAD0492-25G
TCIAD0492-500G
Zinc diethyldithiocarbamate ≥99.0% (by titrimetric analysis)
Zinc diethyldithiocarbamate
Formel:
C₅H₁₁NS₂ |
MDL Number:
MFCD00064798 CAS-nummer: 14324-55-1 EINECS: 238-270-9 UN nummer: 3077 ADR: 9,III |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 99.0 % |
Melting point | 178.0 to 182.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...