2,6-Dichlorophenolindophenol natriumsalt dihydrat ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2,6-Dichloroindophenol, sodium salt dihydrate, Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide dihydrate, Tillmans' reagent dihydrate, 2,6-Dichloroindophenol sodium dihydrate, 2,6-Dichlorophenolindophenol sodium salt, 2,6-Dichloroindophenol sodium salt dihydrate, Sodium 2,6-Dichlorobenzenone-indo-phenol

D0375-1G D0375-25G
TCIAD0375-1GEA 435 DKK
TCIAD0375-1G TCIAD0375-25G
2,6-Dichlorophenolindophenol natriumsalt dihydrat ≥95.0% (by titrimetric analysis)
2,6-Dichlorophenolindophenol natriumsalt hydrat
Formel: C₁₂H₆Cl₂NNaO₂·2H₂O
Molekylvægt: 326.105 g/mol
MDL Number: MFCD00150015
CAS-nummer: 620-45-1
EINECS: 210-640-4
Merck Index: 14,03068
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ed4907bc4b6dcf00f5babf3a67e851d64f4bac&vid=aeff1fbea7211b5cd8702520e527a12f1098a2ad&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Green to Dark green powder to crystal
Purity (Iodometric Titration) min. 95.0 % (calcd.on dried substance)
Drying loss 10.0 to 14.0 %
Interference pigment to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR