Ethylene glycol dibenzyl ether ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D0160-25ML
D0160-5ML
TCIAD0160-25MLEA
1140
DKK
TCIAD0160-25ML
TCIAD0160-5ML
Ethylene glycol dibenzyl ether ≥95.0% (by GC)
Ethylene glycol dibenzyl ether
Formel:
C₁₆H₁₈O₂ Kogepunkt: 210°C |
MDL Number:
MFCD00030016 CAS-nummer: 622-22-0 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...