trans-2-Decenoic acid ≥95.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(E)-2-Decenoic acid, (E)-Dec-2-enoic acid, trans-2-Decylenic Acid
D0098-25ML
D0098-5ML
TCIAD0098-25MLEA
895
DKK
TCIAD0098-25ML
TCIAD0098-5ML
trans-2-Decenoic acid ≥95.0% (by titrimetric analysis)
trans-2-Decenoic acid
Formel:
C₁₀H₁₈O₂ Flammepunkt: 170°C |
MDL Number:
MFCD00039530 CAS-nummer: 334-49-6 EINECS: 206-378-5 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...