2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
2-((6-Chloro-2-methylpyrimidin-4-yl)amino)-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide, N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methyl-4-pyrimidinyl)amino]-5-thiazolecarboxamide
C3675-1G
C3675-5G
TCIAC3675-1GEA
255
DKK
TCIAC3675-1G
TCIAC3675-5G
2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide ≥98.0% (by HPLC)
2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide
Formel:
C₁₆H₁₃Cl₂N₅OS Smeltepunkt: 280°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD11045075 CAS-nummer: 302964-08-5 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...