1-Cyclopentylethanone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C3585-1G C3585-5G
TCIAC3585-1GEA 329 DKK
TCIAC3585-1G TCIAC3585-5G
1-Cyclopentylethanone ≥98.0% (by GC)
1-Cyclopentylethanone
Formel: C₇H₁₂O
Kogepunkt: 156°C
MDL Number: MFCD00060799
CAS-nummer: 6004-60-0
UN nummer: 1224
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR