5-Cyanouracil ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Uracil-5-carbonitrile, 2,4-Dihydroxypyrimidine-5-carbonitrile, 2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile, 5-Cyano-2,4-dioxo-1,2,3,4-tetrahydropyrimidine
C3542-1G
C3542-5G
TCIAC3542-1GEA
591
DKK
TCIAC3542-1G
TCIAC3542-5G
5-Cyanouracil ≥98.0% (by HPLC)
5-Cyanouracil
Formel:
C₅H₃N₃O₂ |
MDL Number:
MFCD00051958 CAS-nummer: 4425-56-3 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...