Clostebol Acetate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Chlorotestosterone Acetate, 17beta-Acetoxy-4-chloro-4-androsten-3-one, 17beta-4-Chloro-3-oxoandrost-4-en-17-yl Acetate, 4-Chloro-17beta-hydroxy-4-androsten-3-one 17-Acetate

C3497-200MG C3497-25MG
TCIAC3497-200MGEA 1990 DKK
TCIAC3497-200MG TCIAC3497-25MG
Clostebol Acetate ≥98.0% (by HPLC, titration analysis)
Clostebol Acetate
Formel: C₂₁H₂₉ClO₃
Molekylvægt: 364.91 g/mol
MDL Number: MFCD00079075
CAS-nummer: 855-19-6
EINECS: 212-720-4
UN nummer: 2811
ADR: 6.1 I
Merck Index: 14,02413

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 227.0 to 231.0 deg-C
Specific rotation (value) 108 and le 119 deg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR