3-Chlorphthalicanhydrid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Chloroisobenzofuran-1,3-dione

C3489-25G C3489-5G
TCIAC3489-25GEA 879 DKK
TCIAC3489-25G TCIAC3489-5G
3-Chlorphthalicanhydrid ≥98.0% (by GC)
3-Chlorphthalicanhydrid
Formel: C₈H₃ClO₃
Molekylvægt: 182.56 g/mol
Kogepunkt: 170°C
MDL Number: MFCD00023107
CAS-nummer: 117-21-5
EINECS: 204-179-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 96.0 %
Melting point 126.0 to 130.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR