N-Carbobenzoxy-1,10-diaminodecane hydrochloride ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
N-Benzyloxycarbonyl-1,10-diaminodecane hydrochloride, Benzyl N-(10-Aminodecyl)carbamate hydrochloride, N-(10-Aminodecyl)carbamic acid Benzyl ester hydrochloride, N-Cbz-1,10-diaminodecane hydrochloride
C3471-1G
C3471-5G
TCIAC3471-1GEA
608
DKK
TCIAC3471-1G
TCIAC3471-5G
N-Carbobenzoxy-1,10-diaminodecane hydrochloride ≥98.0% (by HPLC)
N-Carbobenzoxy-1,10-diaminodecane hydrochloride
Formel:
C₁₈H₃₀N₂O₂·HCl |
MDL Number:
MFCD08272290 CAS-nummer: 1051420-13-3 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 175.0 to 179.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...