CCG-1423 ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-((1-((4-Chlorophenyl)amino)-1-oxopropan-2-yl)oxy)-3,5-bis(trifluoromethyl)benzamide, N-(2-(4-Chloroanilino)-1-methyl-2-oxoethoxy)-3,5-bis(trifluoromethyl)benzamide

C3449-25MG C3449-100MG
TCIAC3449-25MGEA 993 DKK
TCIAC3449-25MG TCIAC3449-100MG
CCG-1423 ≥98.0% (by GC)
CCG-1423
Formel: C18H13ClF6N2O3
Molekylvægt: 454.8 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD01566719
CAS-nummer: 285986-88-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6928b05e38d015754992945db79b9ef3918e8f7&vid=a6928b05e38d015754992945db79b9ef3918e8f7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 5.50 to 6.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR