2-Chloro-1,3-dimethylimidazolium tetrafluoroborate ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Chloro-1,3-dimethylimidazolinium tetrafluoroborate, 2-Chloro-1,3-dimethyl-4,5-dihydroimidazolium tetrafluoroborate, CIB, 2-Chloro-1,3-dimethylimidazolidinium tetrafluoroborate

C3444-1G C3444-5G
TCIAC3444-1GEA 197 DKK
TCIAC3444-1G TCIAC3444-5G
2-Chloro-1,3-dimethylimidazolium tetrafluoroborate ≥98.0% (by HPLC)
2-Chloro-1,3-dimethylimidazolium tetrafluoroborate
Formel: C₅H₁₀ClN₂·BF₄
Molekylvægt: 220.4 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09039291
CAS-nummer: 153433-26-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97ce0b488abe10c98454c1306ee7a0f35e5194d7&vid=97ce0b488abe10c98454c1306ee7a0f35e5194d7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 187.0 to 191.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR