Cucurbitacin B ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (R,E)-6-((2S,8S,9R,10R,13R,14S,16R,17R)-2,16-Dihydroxy-4,4,9,13,14-pentamethyl-3,11-dioxo-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)-6-hydroxy-2-methyl-5-oxohept-3-en-2-yl acetate

C3442-5MG C3442-25MG
TCIAC3442-5MGEA 1240 DKK
TCIAC3442-5MG TCIAC3442-25MG
Cucurbitacin B ≥98.0% (by HPLC)
Cucurbitacin B
Formel: C₃₂H₄₆O₈
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07778083
CAS-nummer: 6199-67-3
UN nummer: 2811
ADR: 6.1,II
Merck Index: 14,02614
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c333f0b506b3db8844194e04db636d704e425c3&vid=7c333f0b506b3db8844194e04db636d704e425c3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Water max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR