2-Chlor-5-methoxyphenylboronsyre (contains varying amounts of Anhydride)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Chloro-5-methoxyphenylboronic Acid, 2-Chloro-5-methoxybenzeneboronic acid
C3432-1G
C3432-5G
TCIAC3432-1GEA
632
DKK
TCIAC3432-1G
TCIAC3432-5G
2-Chlor-5-methoxyphenylboronsyre (contains varying amounts of Anhydride)
2-Chlor-5-methoxyphenylboronsyre
Formel:
C₇H₈BClO₃ Smeltepunkt: 115°C |
MDL Number:
MFCD06659858 CAS-nummer: 89694-46-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | 98.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...