Kobber(I)thiocyanat ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Copper thiocyanate, Copper (I) rhodanide, Cuprous thiocyanate, Copper rhodanide

C3417-250G C3417-25G
TCIAC3417-250GEA 1010 DKK
TCIAC3417-250G TCIAC3417-25G
Kobber(I)thiocyanat ≥98.0% (by titrimetric analysis)
Kobber(I)thiocyanat
Formel: CCuNS
Molekylvægt: 121.63 g/mol
MDL Number: MFCD00010980
CAS-nummer: 1111-67-7
EINECS: 214-183-1
UN nummer: 3077
ADR: 9 III
Merck Index: 14,02669

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Red powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR