2-fluorpyridin-4-carbonitril ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-Cyano-2-fluoropyridine
C3383-5G
C3383-25G
TCIAC3383-5GEA
394
DKK
TCIAC3383-5G
TCIAC3383-25G
2-fluorpyridin-4-carbonitril ≥99.0% (by GC)
2-fluorpyridin-4-carbonitril
Formel:
C₆H₃FN₂ |
MDL Number:
MFCD09027299 CAS-nummer: 3939-14-8 UN nummer: 3439 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 99.0 % |
Melting point | 32.0 to 36.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...