3-Chlor-2-iodtoluen ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Chloro-2-iodo-3-methylbenzene
C3364-5G
C3364-25G
TCIAC3364-5GEA
435
DKK
TCIAC3364-5G
TCIAC3364-25G
3-Chlor-2-iodtoluen ≥95.0% (by GC)
3-Chlor-2-iodtoluen
Formel:
C₇H₆ClI Kogepunkt: 125°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00051756 CAS-nummer: 5100-98-1 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Dark green powder to lump |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...