3'-Chlor-4'-Fluoracetophenon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C3346-5G C3346-25G
TCIAC3346-5GEA 238 DKK
TCIAC3346-5G TCIAC3346-25G
3'-Chlor-4'-Fluoracetophenon ≥98.0% (by GC)
3'-Chlor-4'-Fluoracetophenon
Formel: C₈H₆ClFO
MDL Number: MFCD00042203
CAS-nummer: 2923-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efca6f132e9cb518256745e4edb8af1e8adb103a&vid=efca6f132e9cb518256745e4edb8af1e8adb103a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR