(S)-4-Chloro-3-hydroxybutyronitrile ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C3331-1G
C3331-5G
TCIAC3331-1GEA
132
DKK
TCIAC3331-1G
TCIAC3331-5G
(S)-4-Chloro-3-hydroxybutyronitrile ≥98.0% (by GC)
(S)-4-Chloro-3-hydroxybutyronitrile
Formel:
C₄H₆ClNO Flammepunkt: 111°C |
MDL Number:
MFCD00273363 CAS-nummer: 127913-44-4 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -12.0 to -14.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...