5-Chlorosaligenyl-N,N-diisopropylphosphoramidite ≥95.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
6-Chloro-N,N-diisopropyl-4H-benzo[d][1,3,2]dioxaphosphinin-2-amine
C3326-200MG
C3326-1G
TCIAC3326-200MGEA
649
DKK
TCIAC3326-200MG
TCIAC3326-1G
5-Chlorosaligenyl-N,N-diisopropylphosphoramidite ≥95.0% (by titrimetric analysis)
5-Chlorosaligenyl-N,N-diisopropylphosphoramidite
Formel:
C₁₃H₁₉ClNO₂P |
MDL Number:
MFCD30725743 CAS-nummer: 1620086-77-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...