5-Chlor-α,α,α,2-tetrafluortoluen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C3317-1G C3317-5G
TCIAC3317-1GEA 271 DKK
TCIAC3317-1G TCIAC3317-5G
5-Chlor-α,α,α,2-tetrafluortoluen ≥98.0% (by GC)
5-Chlor-α,α,α,2-tetrafluortoluen
Formel: C₇H₃ClF₄
Flammepunkt: 47°C
MDL Number: MFCD00060655
CAS-nummer: 89634-74-2
UN nummer: 1993
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR