2-Chloro-5-(chloromethyl)thiazole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C3295-25G C3295-5G
TCIAC3295-25GEA 509 DKK
TCIAC3295-25G TCIAC3295-5G
2-Chloro-5-(chloromethyl)thiazole ≥98.0% (by GC)
2-Chloro-5-(chloromethyl)thiazole
Formel: C₄H₃Cl₂NS
Smeltepunkt: 35°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01073549
CAS-nummer: 105827-91-6
EINECS: 429-830-5
UN nummer: 2922
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR