N-(3-Chloro-4-fluorophenyl)-7-fluoro-6-nitro-4-quinazolinamine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

C3276-25G C3276-5G
TCIAC3276-25GEA 1420 DKK
TCIAC3276-25G TCIAC3276-5G
N-(3-Chloro-4-fluorophenyl)-7-fluoro-6-nitro-4-quinazolinamine ≥98.0% (by HPLC, titration analysis)
N-(3-Chloro-4-fluorophenyl)-7-fluoro-6-nitro-4-quinazolinamine
Formel: C₁₄H₇ClF₂N₄O₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09998825
CAS-nummer: 162012-67-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR