Calcium bis(trifluoromethylsulphonyl)imide ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Calcium(II) Bis(trifluoromethanesulphonyl)imide, Calcium bis(trifluoromethanesulphonimide), Bis(trifluoromethanesulphonyl)imide calcium(II) salt, Calcium(II) triflimide, Calcium triflimide

C3263-1G C3263-5G
TCIAC3263-1GEA 386 DKK
TCIAC3263-1G TCIAC3263-5G
Calcium bis(trifluoromethylsulphonyl)imide ≥97.0% (by titrimetric analysis)
Calcium bis(trifluoromethylsulphonyl)imide
Formel: C₄CaF₁₂N₂O₈S₄
Molekylvægt: 600.35 g/mol
MDL Number: MFCD14156019
CAS-nummer: 165324-09-4
UN nummer: 1759
ADR: 8 II

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Purity (Ion exchange titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR