3-Chlor-2-methoxybenzoesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C3261-1G C3261-5G
TCIAC3261-1GEA 501 DKK
TCIAC3261-1G TCIAC3261-5G
3-Chlor-2-methoxybenzoesyre ≥98.0% (by GC, titration analysis)
3-Chlor-2-methoxybenzoesyre
Formel: C₈H₇ClO₃
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00179306
CAS-nummer: 3260-93-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfe61a5b7f464760082274e364651478192a3a15&vid=bfe61a5b7f464760082274e364651478192a3a15&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR