(Chloromethyl)diethoxy(methyl)silane ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Chloromethylmethyldiethoxysilane

C3257-5G C3257-25G
TCIAC3257-5GEA 312 DKK
TCIAC3257-5G TCIAC3257-25G
(Chloromethyl)diethoxy(methyl)silane ≥98.0% (by GC)
(Chloromethyl)diethoxy(methyl)silane
Formel: C₆H₁₅ClO₂Si
Molekylvægt: 182.72 g/mol
Kogepunkt: 161°C
Flammepunkt: 37°C
MDL Number: MFCD00018878
CAS-nummer: 2212-10-4
EINECS: 218-657-9
UN nummer: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26ad640befe4042dbd482ad0e6722830bf24851b&vid=26ad640befe4042dbd482ad0e6722830bf24851b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR