4-Chlor-α,α,α,3-tetrafluortoluen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C3216-1G C3216-5G
TCIAC3216-1GEA 230 DKK
TCIAC3216-1G TCIAC3216-5G
4-Chlor-α,α,α,3-tetrafluortoluen ≥98.0% (by GC)
4-Chlor-α,α,α,3-tetrafluortoluen
Formel: C₇H₃ClF₄
MDL Number: MFCD00060646
CAS-nummer: 32137-20-5
UN nummer: 1993
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR