ω-Chloro-3,4-difluoroacetophenone ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Chloro-3',4'-difluoroacetophenone, 2-Chloro-1-(3,4-difluorophenyl)ethanone
C3215-1G
C3215-5G
TCIAC3215-1GEA
238
DKK
TCIAC3215-1G
TCIAC3215-5G
ω-Chloro-3,4-difluoroacetophenone ≥98.0% (by GC)
ω-Chloro-3,4-difluoroacetophenone
Formel:
C₈H₅ClF₂O Molekylvægt: 190.5745 g/mol |
MDL Number:
MFCD03966888 CAS-nummer: 51336-95-9 UN nummer: 3261 ADR: 8 II |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...