Chloroformamidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Carbamimidic chloride hydrochloride

C3199-5G C3199-25G
TCIAC3199-5GEA 181 DKK
TCIAC3199-5G TCIAC3199-25G
Chloroformamidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Chloroformamidine hydrochloride
Formel: CH₄Cl₂N₂
Smeltepunkt: 180°C
MDL Number: MFCD00035527
CAS-nummer: 29671-92-9
EINECS: 249-765-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR