5-Chloro-1-methylisatin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Chloro-1-methylindole-2,3-dione, 5-Chloro-1-methylindoline-2,3-dione, 5-Chloro-1-methyl-2,3-indolinedione
C3195-200MG
C3195-1G
TCIAC3195-200MGEA
559
DKK
TCIAC3195-200MG
TCIAC3195-1G
5-Chloro-1-methylisatin ≥98.0% (by GC)
5-Chloro-1-methylisatin
Formel:
C₉H₆ClNO₂ Molekylvægt: 195.60 g/mol |
MDL Number:
MFCD02956479 CAS-nummer: 60434-13-1 EINECS: 262-231-5 |
Specifikationer Testresultater
Appearance | Orange to Red powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 175.0 to 179.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...