3-Chlor-4-methylanisol ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Chloro-4-methoxy-1-methylbenzene, 3-Chloro-4-methylanisole, 2-Chloro-4-methoxytoluene

C3190-5G C3190-25G
TCIAC3190-5GEA 411 DKK
TCIAC3190-5G TCIAC3190-25G
3-Chlor-4-methylanisol ≥95.0% (by GC)
3-Chlor-4-methylanisol
Formel: C₈H₉ClO
Kogepunkt: 213°C
MDL Number: MFCD00087376
CAS-nummer: 54788-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e005db93db51ee33f4497ef4bfe570f6256a2ed4&vid=e005db93db51ee33f4497ef4bfe570f6256a2ed4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR