N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
N-(4-Cyano-3-(trifluoromethyl)phenyl)methacrylamide
C3188-1G
C3188-5G
TCIAC3188-1GEA
476
DKK
TCIAC3188-1G
TCIAC3188-5G
N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide ≥98.0% (by GC)
N-[4-Cyano-3-(trifluoromethyl)phenyl]methacrylamide
Formel:
C₁₂H₉F₃N₂O Molekylvægt: 254.21 g/mol |
MDL Number:
MFCD03411609 CAS-nummer: 90357-53-2 EINECS: 427-880-2 UN nummer: 3077 ADR: 9 III |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 141.0 to 145.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...