Cyclopentanecarbonitrile ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C3167-5ML C3167-25ML
TCIAC3167-5MLEA 846 DKK
TCIAC3167-5ML TCIAC3167-25ML
Cyclopentanecarbonitrile ≥98.0% (by GC)
Cyclopentanecarbonitrile
Formel: C₆H₉N
Kogepunkt: 168°C
Flammepunkt: 56°C
MDL Number: MFCD00013741
CAS-nummer: 4254-02-8
EINECS: 224-229-2
UN nummer: 1992
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR