3-Chlorsalicylaldehyd ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Chloro-2-hydroxybenzaldehyde, 2-Hydroxy-3-chlorobenzaldehyde

C3166-1G C3166-5G
TCIAC3166-1GEA 403 DKK
TCIAC3166-1G TCIAC3166-5G
3-Chlorsalicylaldehyd ≥98.0% (by GC, titration analysis)
3-Chlorsalicylaldehyd
Formel: C₇H₅ClO₂
Kogepunkt: 63°C
MDL Number: MFCD04973581
CAS-nummer: 1927-94-2

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR