2-Chlor-4-(trifluormethoxy)anilin ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C3142-1G C3142-5G
TCIAC3142-1GEA 189 DKK
TCIAC3142-1G TCIAC3142-5G
2-Chlor-4-(trifluormethoxy)anilin ≥97.0% (by GC, titration analysis)
2-Chlor-4-(trifluormethoxy)anilin
Formel: C₇H₅ClF₃NO
Molekylvægt: 211.57 g/mol
Flammepunkt: 98°C
MDL Number: MFCD01631704
CAS-nummer: 69695-61-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR